Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000463413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000463413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000463413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000463413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000495724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
ENST00000495724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012680.hg.1 |
TC07000805.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012680.hg.1 |
ENST00000335420.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012680.hg.1 |
ENST00000495724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012680.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164878.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164878.hg.1 |
ENST00000463413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164878.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164878.hg.1 |
ENST00000495724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164878.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164878.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164878.hg.1 |
ENST00000463413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164878.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164878.hg.1 |
ENST00000495724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164878.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164878.hg.1 |
TC0700009122.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000335420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000463413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000463413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000463413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000463413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000468226.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000495724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
ENST00000495724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009122.hg.1 |
NM_014997.3 |
Refseq |