Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002334.3 | NP_002325.2 | 35 - 36 | 5392 | 64 | |
XM_011520103.2 | XP_011518405.1 | 29 - 30 | 4462 | 64 | ||
XM_011520104.2 | XP_011518406.1 | 20 - 21 | 2929 | 64 | ||
XM_017017734.1 | XP_016873223.1 | 35 - 36 | 5309 | 64 | ||
GenBank mRNA | AB011540.1 | - | 27 - 28 | 4162 | 64 | |
AB084910.1 | - | 35 - 36 | 5392 | 64 | ||
AB231861.1 | - | 35 - 36 | 5286 | 64 | ||
BC136667.1 | - | 35 - 36 | 5313 | 9020657 | 64 | |
BC136668.1 | - | 35 - 36 | 5313 | 9020658 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |