Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
ENST00000338957.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011300.hg.1 |
NM_001010888.3 |
Refseq |