Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011132.hg.1 |
TC08000841.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011132.hg.1 |
ENST00000322428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011132.hg.1 |
ENST00000532522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011132.hg.1 |
ENST00000534585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011132.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000527572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009240.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163111.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163111.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163111.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163111.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163111.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163111.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163111.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163111.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163111.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163111.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163111.hg.1 |
TC0800009240.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
ENST00000322428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
ENST00000527058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
ENST00000532522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
ENST00000534585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
ENST00000534811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
NM_032272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163110.hg.1 |
TC0800009240.hg.1 |
Clariom D TC |