Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006988.hg.1 |
ENST00000163416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006988.hg.1 |
ENST00000439315.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006988.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006988.hg.1 |
TC14000572.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006987.hg.1 |
ENST00000163416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006987.hg.1 |
ENST00000355976.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006987.hg.1 |
ENST00000439315.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006987.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006987.hg.1 |
TC14000572.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000554700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000554700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000554700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000554700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000555793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000555793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
ENST00000555793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000554700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000554700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000554700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000554700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000555793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000555793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
ENST00000555793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008019.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400156778.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400156778.hg.1 |
ENST00000554700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400156778.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400156778.hg.1 |
ENST00000163416.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400156778.hg.1 |
ENST00000554700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400156778.hg.1 |
NM_005113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400156778.hg.1 |
TC1400008019.hg.1 |
Clariom D TC |