Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011257.hg.1 |
TC06000963.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011257.hg.1 |
ENST00000229633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011257.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009368.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164281.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164281.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164281.hg.1 |
ENST00000229633.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164281.hg.1 |
NM_138571.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164281.hg.1 |
TC0600009368.hg.1 |
Clariom D TC |