Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000475028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000475028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
ENST00000475028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170691.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170691.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170691.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170691.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170691.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170691.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170691.hg.1 |
TC0600010375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000230449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000443083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000475028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000475028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
ENST00000475028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NM_018303.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010375.hg.1 |
NR_073064.1 |
Refseq |