Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
ENST00000491428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
ENST00000491428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
ENST00000491428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028156.hg.1 |
TC01001804.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028156.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028156.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
ENST00000491428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
ENST00000491428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
ENST00000491428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011619.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185536.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185536.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185536.hg.1 |
ENST00000366932.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185536.hg.1 |
NM_016052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185536.hg.1 |
TC0100011619.hg.1 |
Clariom D TC |