Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007517.hg.1 |
ENST00000164133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007517.hg.1 |
ENST00000359279.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007517.hg.1 |
ENST00000527441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007517.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007517.hg.1 |
TC11000611.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100201145.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100201145.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100201145.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100201145.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100201145.hg.1 |
TC1100013036.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |