Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000261180.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000547300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000547300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000547300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000547300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000547300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000549922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000552503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000552503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000552503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000552503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
ENST00000552503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012668.hg.1 |
NM_013381.2 |
Refseq |