Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005691.3 | NP_005682.2 | 11 - 12 | 1675 | 85 | |
NM_020297.3 | NP_064693.2 | 11 - 12 | 1675 | 85 | ||
XM_005253284.3 | XP_005253341.1 | 13 - 14 | 2461 | 85 | ||
XM_005253286.3 | XP_005253343.1 | 14 - 15 | 2521 | 85 | ||
XM_005253287.4 | XP_005253344.1 | 13 - 14 | 2461 | 85 | ||
XM_005253288.3 | XP_005253345.1 | 12 - 13 | 2345 | 85 | ||
XM_005253289.3 | XP_005253346.1 | 13 - 14 | 2461 | 85 | ||
XM_005253290.3 | XP_005253347.1 | 13 - 14 | 2461 | 85 | ||
XM_006719025.3 | XP_006719088.1 | 13 - 14 | 2461 | 85 | ||
XM_011520545.2 | XP_011518847.1 | 13 - 14 | 2517 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |