Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013962.hg.1 |
TC08001087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013962.hg.1 |
ENST00000301906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013962.hg.1 |
ENST00000354914.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013962.hg.1 |
ENST00000380385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013962.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013962.hg.1 |
ENST00000524352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013962.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013964.hg.1 |
TC08001087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013964.hg.1 |
ENST00000301906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013964.hg.1 |
ENST00000354914.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013964.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013964.hg.1 |
ENST00000524352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013964.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013963.hg.1 |
TC08001087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013963.hg.1 |
ENST00000301906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013963.hg.1 |
ENST00000354914.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013963.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013963.hg.1 |
ENST00000524352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013963.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167820.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167820.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167820.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167820.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167820.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167820.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167820.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167820.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167820.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167820.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167820.hg.1 |
TC0800009971.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000354914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000380385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000518030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
ENST00000524352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009971.hg.1 |
NM_173833.5 |
Refseq |