Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001898.hg.1 |
TC6_mann_hap4000115.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001898.hg.1 |
NM_002714.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002378.hg.1 |
TC6_cox_hap2000137.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002378.hg.1 |
NM_002714.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002308.hg.1 |
TC6_dbb_hap3000126.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002308.hg.1 |
NM_002714.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016937.hg.1 |
TC06001502.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016937.hg.1 |
ENST00000376511.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016937.hg.1 |
NM_002714.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002068.hg.1 |
TC6_mcf_hap5000116.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002068.hg.1 |
NM_002714.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002351.hg.1 |
TC6_qbl_hap6000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002351.hg.1 |
NM_002714.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002152.hg.1 |
TC6_ssto_hap7000118.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002152.hg.1 |
NM_002714.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176625.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176625.hg.1 |
ENST00000376511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176625.hg.1 |
TC0600011372.hg.1 |
Clariom D TC |