Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024354.hg.1 |
TC06001968.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024354.hg.1 |
ENST00000281806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024354.hg.1 |
ENST00000369212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024354.hg.1 |
ENST00000445970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024354.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024354.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012664.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187347.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187347.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187347.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187347.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187347.hg.1 |
ENST00000281806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187347.hg.1 |
ENST00000369212.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187347.hg.1 |
NM_001040179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187347.hg.1 |
NM_032503.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187347.hg.1 |
TC0600012664.hg.1 |
Clariom D TC |