Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000465018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000465018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000465018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005260.hg.1 |
ENST00000255305.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005260.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005260.hg.1 |
ENST00000456108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005260.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005260.hg.1 |
TC13000461.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000465018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000465018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000465018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
ENST00000490513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008267.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155816.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155816.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155816.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155816.hg.1 |
ENST00000255305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155816.hg.1 |
ENST00000400602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155816.hg.1 |
NM_022459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155816.hg.1 |
TC1300008267.hg.1 |
Clariom D TC |