Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016919.hg.1 |
ENST00000302824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016919.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016919.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016919.hg.1 |
TC15001730.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185711.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185711.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185711.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185711.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185711.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185711.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185711.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185711.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185711.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185711.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185711.hg.1 |
TC1500010909.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000325346.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000559913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
ENST00000560723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010909.hg.1 |
NM_181900.2 |
Refseq |