Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017586.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017586.hg.1 |
ENST00000561140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017586.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017586.hg.1 |
TC15001816.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017587.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017587.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017587.hg.1 |
TC15001816.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000559538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000559538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000559538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000559538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000560703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000560703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000560703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
ENST00000560703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185993.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185993.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185993.hg.1 |
ENST00000560703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185993.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185993.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185993.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185993.hg.1 |
ENST00000560703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185993.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185993.hg.1 |
TC1500010921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500185991.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185991.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185991.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185991.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185991.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185991.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185991.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185991.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185991.hg.1 |
TC1500010921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000558660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000559538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000559538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000559538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000559538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000560703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000560703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000560703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
ENST00000560703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010921.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185992.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185992.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185992.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185992.hg.1 |
ENST00000312475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185992.hg.1 |
ENST00000558531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185992.hg.1 |
NM_022163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185992.hg.1 |
TC1500010921.hg.1 |
Clariom D TC |