Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318391.1 | NP_001305320.1 | 12 - 13 | 1484 | 75 | |
NM_032830.2 | NP_116219.2 | 12 - 13 | 1538 | 75 | ||
XM_005256205.3 | XP_005256262.2 | 8 - 9 | 1069 | 75 | ||
GenBank mRNA | AB075868.2 | - | 12 - 13 | 1476 | 75 | |
AK027419.1 | - | 9 - 10 | 1093 | 75 | ||
AK027584.1 | - | 1 - 2 | 115 | 75 | ||
AK027634.1 | - | 12 - 13 | 1490 | 75 | ||
AK027675.1 | - | 12 - 13 | 1487 | 75 | ||
AK074795.1 | - | 12 - 13 | 1504 | 75 | ||
BC000167.2 | - | 8 - 9 | 983 | 75 | ||
BC009348.2 | - | 12 - 13 | 1470 | 4131625 | 75 | |
BX647265.1 | - | 12 - 13 | 1452 | 75 | ||
CA430627.1 | - | 3 - 4 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |