Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012646.hg.1 |
ENST00000216200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012646.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012646.hg.1 |
TC22000715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012642.hg.1 |
ENST00000216200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012642.hg.1 |
ENST00000417718.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012642.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012642.hg.1 |
TC22000715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012640.hg.1 |
ENST00000216200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012640.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012640.hg.1 |
ENST00000417718.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012640.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012640.hg.1 |
TC22000715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000443735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000443735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000443735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000443735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000467935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000467935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000467935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
ENST00000467935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
TC2200008621.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000443735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000467935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000443735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
ENST00000467935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162187.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000216200.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000404171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000406910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000417718.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000443735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000443735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000443735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000443735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000467935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000467935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000467935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
ENST00000467935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008621.hg.1 |
NM_002854.2 |
Refseq |