Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
ENST00000052754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
ENST00000228329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
ENST00000393155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
ENST00000420120.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
ENST00000425043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
ENST00000547568.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
ENST00000552962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
NM_001920.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
NM_133503.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
NM_133504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
NM_133505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023851.hg.1 |
TC12001804.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000052754.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000393155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000420120.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000425043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000546391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000547568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000550758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000552962.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
NM_001920.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
NM_133503.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
NM_133504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
NM_133505.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000052754.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000393155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000420120.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000425043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000546391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000547568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000550758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
ENST00000552962.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
NM_001920.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
NM_133503.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
NM_133504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
NM_133505.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189081.hg.1 |
TC1200011496.hg.1 |
Clariom D TC |