Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NALCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052867.2 | NP_443099.1 | 39 - 40 | 4632 | 117 | |
XM_011521067.2 | XP_011519369.1 | 39 - 40 | 4572 | 117 | ||
XM_011521068.2 | XP_011519370.1 | 39 - 40 | 4555 | 117 | ||
XM_011521069.2 | XP_011519371.1 | 38 - 39 | 4485 | 117 | ||
XM_017020534.1 | XP_016876023.1 | 38 - 39 | 4516 | 117 | ||
XM_017020535.1 | XP_016876024.1 | 38 - 39 | 4458 | 117 | ||
XM_017020536.1 | XP_016876025.1 | 34 - 35 | 4384 | 117 | ||
XM_017020537.1 | XP_016876026.1 | 33 - 34 | 3773 | 117 | ||
GenBank mRNA | AK055506.1 | - | 6 - 7 | 689 | 117 | |
AK126621.1 | - | 20 - 21 | 2096 | 117 | ||
AK126718.1 | - | 12 - 13 | 1233 | 117 | ||
AK172752.1 | - | 2 - 3 | 163 | 117 | ||
AY141972.1 | - | 39 - 40 | 4526 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Homo sapiens Probe spans exons |