Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK293934.1 | - | 1 - 2 | 89 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |