Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006881.hg.1 |
ENST00000595307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006881.hg.1 |
ENST00000595307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006881.hg.1 |
ENST00000595307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006881.hg.1 |
ENST00000595307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000616513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
ENST00000616513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002618.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002618.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002618.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002618.hg.1 |
TC13000220.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300149103.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149103.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149103.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149103.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149103.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149103.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149103.hg.1 |
TC1300007237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1900006881.hg.1 |
ENST00000595307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006881.hg.1 |
ENST00000595307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006881.hg.1 |
ENST00000595307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006881.hg.1 |
ENST00000595307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000519151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000521776.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000523764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000616513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
ENST00000616513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_001004127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NM_021645.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007237.hg.1 |
NR_036571.2 |
Refseq |