Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000396901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000585925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000586361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588497.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588497.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588497.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588497.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588497.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588497.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000588497.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000590828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000591730.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000591730.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000591730.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000591730.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000591730.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000591730.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000591730.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000591730.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000606253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000624076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000624076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
ENST00000624076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010502.hg.1 |
NM_139239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000465810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000465810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000465810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000465810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000465810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000340171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000465810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000465810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000465810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000465810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000465810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000482061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000486435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
ENST00000534941.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_001308229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012880.hg.1 |
NM_173657.2 |
Refseq |