Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005720.hg.1 |
TC09000519.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005720.hg.1 |
ENST00000389120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005720.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000478072.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000478072.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000478072.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000478072.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000479306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000479306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000479306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000479306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000479306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000481046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000481046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000481046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000481046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
ENST00000481046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900155346.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900155346.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900155346.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900155346.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900155346.hg.1 |
TC0900008257.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000374819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000389120.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000466817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000478072.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000478072.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000478072.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000478072.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000479306.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000479306.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000479306.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000479306.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000479306.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000481046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000481046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000481046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000481046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
ENST00000481046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008257.hg.1 |
NM_019592.6 |
Refseq |