Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128427.2 | NP_001121899.1 | 2 - 3 | 554 | 96 | |
NM_001128428.2 | NP_001121900.1 | 2 - 3 | 554 | 96 | ||
GenBank mRNA | AB033089.1 | - | 2 - 3 | 484 | 96 | |
AK315152.1 | - | 2 - 3 | 525 | 96 | ||
BC073978.1 | - | 2 - 3 | 257 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |