Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030750.hg.1 |
TC03001699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030750.hg.1 |
ENST00000310864.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030750.hg.1 |
ENST00000349820.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030750.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030750.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030758.hg.1 |
TC03001699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030758.hg.1 |
ENST00000310864.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030758.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030758.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030751.hg.1 |
TC03001699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030751.hg.1 |
ENST00000310864.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030751.hg.1 |
ENST00000349820.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030751.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030751.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190692.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190692.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190692.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190692.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190692.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190692.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190692.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190692.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190692.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190692.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190692.hg.1 |
TC0300012171.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190686.hg.1 |
TC0300012171.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190687.hg.1 |
TC0300012171.hg.1 |
Clariom D TC |