Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001910.hg.1 |
TC06000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001910.hg.1 |
ENST00000465856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001910.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000463944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000463944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000463944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000463944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000463944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000475151.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000475151.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000475151.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000475151.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
ENST00000475151.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001909.hg.1 |
TC06000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001909.hg.1 |
ENST00000465856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001909.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001906.hg.1 |
TC06000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001906.hg.1 |
ENST00000465856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001906.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001907.hg.1 |
TC06000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001907.hg.1 |
ENST00000465856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001907.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001908.hg.1 |
TC06000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001908.hg.1 |
ENST00000465856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001908.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149557.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149557.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149557.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149557.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149557.hg.1 |
TC0600007305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000463944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000463944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000463944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000463944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000463944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000475151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000475151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000475151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000475151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
ENST00000475151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007305.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149558.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149558.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149558.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149558.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149558.hg.1 |
TC0600007305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600149561.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149561.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149561.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149561.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149561.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149561.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149561.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149561.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149561.hg.1 |
TC0600007305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600149562.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149562.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149562.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149562.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149562.hg.1 |
TC0600007305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600149560.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149560.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149560.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149560.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149560.hg.1 |
ENST00000377662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149560.hg.1 |
ENST00000465856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149560.hg.1 |
ENST00000466808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149560.hg.1 |
NR_027795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149560.hg.1 |
TC0600007305.hg.1 |
Clariom D TC |