Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291860.1 | NP_001278789.1 | 43 - 44 | 5482 | 90 | |
NM_005529.6 | NP_005520.4 | 43 - 44 | 5479 | 90 | ||
XM_011541318.2 | XP_011539620.1 | 48 - 49 | 5963 | 90 | ||
XM_017001120.1 | XP_016856609.1 | 44 - 45 | 5609 | 90 | ||
XM_017001121.1 | XP_016856610.1 | 43 - 44 | 5558 | 90 | ||
XM_017001122.1 | XP_016856611.1 | 43 - 44 | 5555 | 90 | ||
GenBank mRNA | M85289.1 | - | 43 - 44 | 5479 | 90 | |
X62515.1 | - | 43 - 44 | 5445 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons |