Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291860.1 | NP_001278789.1 | 61 - 62 | 8104 | 96 | |
NM_005529.6 | NP_005520.4 | 61 - 62 | 8101 | 96 | ||
XM_011541318.2 | XP_011539620.1 | 66 - 67 | 8585 | 96 | ||
XM_017001120.1 | XP_016856609.1 | 62 - 63 | 8231 | 96 | ||
XM_017001121.1 | XP_016856610.1 | 61 - 62 | 8180 | 96 | ||
XM_017001122.1 | XP_016856611.1 | 61 - 62 | 8177 | 96 | ||
GenBank mRNA | AB209851.1 | - | 13 - 14 | 1841 | 96 | |
M85289.1 | - | 61 - 62 | 8101 | 96 | ||
X62515.1 | - | 61 - 62 | 8067 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons |