Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003811.1 | NP_001003811.1 | 22 - 23 | 2005 | 66 | |
NM_031276.2 | NP_112566.2 | 21 - 22 | 1883 | 66 | ||
XM_011530994.1 | XP_011529296.1 | 21 - 22 | 1877 | 66 | ||
XM_011530996.1 | XP_011529298.1 | 10 - 11 | 1023 | 66 | ||
XM_017029649.1 | XP_016885138.1 | 21 - 22 | 1877 | 66 | ||
XM_017029650.1 | XP_016885139.1 | 10 - 11 | 1069 | 66 | ||
XM_017029651.1 | XP_016885140.1 | 10 - 11 | 1023 | 66 | ||
XM_017029652.1 | XP_016885141.1 | 8 - 9 | 704 | 66 | ||
GenBank mRNA | AF285594.1 | - | 20 - 21 | 1788 | 66 | |
AK057523.1 | - | 22 - 23 | 1999 | 66 | ||
AK292471.1 | - | 21 - 22 | 1877 | 66 | ||
BC036016.1 | - | 21 - 22 | 1883 | 5270970 | 66 | |
BU929507.1 | - | 2 - 3 | 138 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |