Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000356.hg.1 |
TC05000017.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000356.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000356.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000353.hg.1 |
TC05000017.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000353.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000353.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000354.hg.1 |
TC05000017.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000354.hg.1 |
ENST00000296821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000354.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000354.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000513607.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000513607.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000513607.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
ENST00000513607.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000513607.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000513607.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000513607.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
ENST00000513607.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006494.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144692.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144692.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144692.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144692.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144692.hg.1 |
TC0500006494.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500144693.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144693.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144693.hg.1 |
ENST00000324642.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144693.hg.1 |
NM_182632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144693.hg.1 |
TC0500006494.hg.1 |
Clariom D TC |