Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143994.1 | NP_001137466.1 | 4 - 5 | 982 | 108 | |
XM_005253149.3 | XP_005253206.2 | 5 - 6 | 1133 | 108 | ||
XM_017018362.1 | XP_016873851.1 | 4 - 5 | 1044 | 108 | ||
GenBank mRNA | BC106922.1 | - | 4 - 5 | 990 | 40032778 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |