Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153161.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153161.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153161.hg.1 |
TC2100007881.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153161.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153161.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153162.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153162.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153162.hg.1 |
TC2100007881.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153162.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153162.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153164.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153164.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153164.hg.1 |
TC2100007881.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153164.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153164.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
ENST00000399899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007881.hg.1 |
NM_199328.2 |
Refseq |