Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007668.hg.1 |
ENST00000229088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007668.hg.1 |
ENST00000411580.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007668.hg.1 |
ENST00000455166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007668.hg.1 |
ENST00000539867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007668.hg.1 |
ENST00000542120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007668.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007668.hg.1 |
TC12000576.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000456757.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000456757.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000456757.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000456757.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000456757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000456757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000456757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000456757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008016.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
ENST00000539120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
ENST00000539867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
ENST00000542120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
ENST00000544190.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
ENST00000544457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
NM_015279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158742.hg.1 |
TC1200008016.hg.1 |
Clariom D TC |