Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017028024.1 | XP_016883513.1 | 7 - 8 | 1090 | 113 | |
XM_017028025.1 | XP_016883514.1 | 6 - 7 | 971 | 113 | ||
XM_017028026.1 | XP_016883515.1 | 6 - 7 | 810 | 113 | ||
XM_017028027.1 | XP_016883516.1 | 4 - 5 | 387 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |