Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001283.hg.1 |
ENST00000380071.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001283.hg.1 |
ENST00000434425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001283.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001283.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001283.hg.1 |
TC13000119.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300146578.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146578.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146578.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146578.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146578.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146578.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146578.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146578.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146578.hg.1 |
TC1300006847.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |