Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000473079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000473079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000473079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000482987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000482987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000482987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
ENST00000482987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03017767.hg.1 |
TC03001007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03017767.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03017767.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300170546.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300170546.hg.1 |
ENST00000482987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300170546.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300170546.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300170546.hg.1 |
ENST00000482987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300170546.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300170546.hg.1 |
TC0300009713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000330394.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000473079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000473079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000473079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000482987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000482987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000482987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
ENST00000482987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009713.hg.1 |
NM_004443.3 |
Refseq |