Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007988.hg.1 |
TC05000573.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007988.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007988.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000504881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000504881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
ENST00000504881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000504881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000504881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
ENST00000504881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008467.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158063.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158063.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158063.hg.1 |
ENST00000339397.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158063.hg.1 |
NM_152546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158063.hg.1 |
TC0500008467.hg.1 |
Clariom D TC |