Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031714.3 | NP_001026884.3 | 4 - 5 | 814 | 83 | |
NM_022489.3 | NP_071934.3 | 4 - 5 | 814 | 83 | ||
XM_005268004.4 | XP_005268061.1 | 4 - 5 | 901 | 83 | ||
XM_005268005.4 | XP_005268062.1 | 4 - 5 | 872 | 83 | ||
XM_017021595.1 | XP_016877084.1 | 4 - 5 | 872 | 83 | ||
GenBank mRNA | BQ961756.1 | - | 1 - 2 | 100 | 83 | |
BX248757.1 | - | 4 - 5 | 783 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |