Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011723.hg.1 |
TC06000994.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011723.hg.1 |
ENST00000275198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011723.hg.1 |
NM_175067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009479.hg.1 |
ENST00000275198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009479.hg.1 |
NM_175067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011724.hg.1 |
TC06000994.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011724.hg.1 |
ENST00000275198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011724.hg.1 |
ENST00000539228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011724.hg.1 |
NM_175067.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009479.hg.1 |
ENST00000275198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009479.hg.1 |
NM_175067.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165051.hg.1 |
ENST00000275198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165051.hg.1 |
NM_175067.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165051.hg.1 |
ENST00000275198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165051.hg.1 |
NM_175067.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165051.hg.1 |
TC0600009479.hg.1 |
Clariom D TC |