Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000297792.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000449850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
ENST00000546309.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007103.hg.1 |
NM_153042.3 |
Refseq |