Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
TC02005011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
ENST00000373563.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
ENST00000373566.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
ENST00000409196.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
ENST00000409451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
ENST00000409480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
ENST00000409547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
ENST00000414511.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
ENST00000535418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
NM_001103146.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
NM_001103147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
NM_001103148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022314.hg.1 |
NM_015575.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
TC02005011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
ENST00000373563.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
ENST00000373566.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
ENST00000409196.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
ENST00000409451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
ENST00000409480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
ENST00000409547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
ENST00000414511.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
ENST00000535418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
NM_001103146.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
NM_001103147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
NM_001103148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022315.hg.1 |
NM_015575.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
TC02005011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
ENST00000373563.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
ENST00000373566.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
ENST00000409196.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
ENST00000409451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
ENST00000409480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
ENST00000409547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
ENST00000414511.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
ENST00000535418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
NM_001103146.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
NM_001103147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
NM_001103148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022312.hg.1 |
NM_015575.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
TC02005011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
ENST00000373563.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
ENST00000373566.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
ENST00000409196.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
ENST00000409451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
ENST00000409480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
ENST00000409547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
ENST00000414511.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
ENST00000535418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
NM_001103146.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
NM_001103147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
NM_001103148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022313.hg.1 |
NM_015575.3 |
Refseq |