Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010734.hg.1 |
TC08000813.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010734.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010734.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010738.hg.1 |
TC08000813.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010738.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010738.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009166.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162343.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162343.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162343.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162343.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162343.hg.1 |
ENST00000522728.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162343.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162343.hg.1 |
TC0800009166.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800162340.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162340.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162340.hg.1 |
ENST00000220940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162340.hg.1 |
NM_002066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162340.hg.1 |
TC0800009166.hg.1 |
Clariom D TC |