Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015061.hg.1 |
TC0X001103.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015061.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015061.hg.1 |
ENST00000540071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015061.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000426382.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000484842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000484842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000484842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000484842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000484842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000486068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000491714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000491714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
ENST00000491714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000426382.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000467444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000484842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000484842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000484842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000484842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000484842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000486068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000491714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000491714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
ENST00000491714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009929.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167833.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00167833.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167833.hg.1 |
ENST00000355520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00167833.hg.1 |
NM_002547.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167833.hg.1 |
TC0X00009929.hg.1 |
Clariom D TC |