Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001402.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001402.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001402.hg.1 |
TC11000082.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001400.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001400.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001400.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001400.hg.1 |
TC11000082.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001398.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001398.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001398.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001398.hg.1 |
TC11000082.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001401.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001401.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001401.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001401.hg.1 |
TC11000082.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100146373.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146373.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146373.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146373.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146373.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146373.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146373.hg.1 |
TC1100006629.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006629.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146371.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146371.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146371.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146371.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146371.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146371.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146371.hg.1 |
TC1100006629.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100146375.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146375.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146375.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146375.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146375.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146375.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146375.hg.1 |
TC1100006629.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100146376.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146376.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146376.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146376.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146376.hg.1 |
TC1100006629.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100146374.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146374.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146374.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146374.hg.1 |
ENST00000250693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146374.hg.1 |
ENST00000529556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146374.hg.1 |
NM_004314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146374.hg.1 |
TC1100006629.hg.1 |
Clariom D TC |