Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000494326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000494326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000494326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000494326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036343.hg.1 |
TC02002254.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036343.hg.1 |
ENST00000019103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036343.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202796.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202796.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202796.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202796.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202796.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202796.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202796.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202796.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202796.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202796.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202796.hg.1 |
TC0200014034.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000019103.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000485440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000494326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000494326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000494326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000494326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000627145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
ENST00000630739.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014034.hg.1 |
NM_002980.2 |
Refseq |