Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000310864.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000349820.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000393650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000460108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000462442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000471726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
ENST00000498104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012171.hg.1 |
NM_001023571.3 |
Refseq |