Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000506168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000506168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000506168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000506168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003390.hg.1 |
TC05000275.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003390.hg.1 |
ENST00000381070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003390.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003390.hg.1 |
ENST00000538793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003390.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000506168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000506168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000506168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000506168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
ENST00000545000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007591.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
ENST00000381070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
ENST00000485990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
ENST00000508024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
NM_001297644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
NM_001297645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
NM_005869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151210.hg.1 |
TC0500007591.hg.1 |
Clariom D TC |