Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003239.3 | NP_003230.1 | 2 - 3 | 1645 | 68 | |
GenBank mRNA | AK313564.1 | - | 2 - 3 | 726 | 68 | |
BC018503.1 | - | 2 - 3 | 616 | 4477175 | 68 | |
BT007287.1 | - | 2 - 3 | 516 | 68 | ||
EF560714.1 | - | 2 - 3 | 629 | 68 | ||
J03241.1 | - | 2 - 3 | 778 | 68 | ||
X14149.1 | - | 2 - 3 | 769 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |